Learn More
Electron Microscopy Sciences SEM Carbon Planchet, 25.4 mm, 1.6 mm height
SDP
Supplier:  Electron Microscopy Sciences 76272
Encompass_Preferred
Carbon planchet for scanning electron microscopy (SEM).
Surface: 1"
Height: 1.6 mm
Catalog No. 50-248-96