Learn More
TELEDYNE CETAC TECHNOLOGIES Sample Probes for XLR-8 Auto Sampler, 0.8 OD, Red
SDP
Catalog No. NC0417035
Encompass
Length: 10 inches (25.4 cm), Probe Material: FEP-covered carbon fiber, Capillary Material: PFA, Capillary Length: 108 in (2.74 m)
Catalog No. NC0417035 Supplier TELEDYNE CETAC TECHNOLOGIES Supplier No. SP5796C
Length: 10 inches (25.4 cm), Probe Material: FEP-covered carbon fiber, Capillary Material: PFA, Capillary Length: 108 in (2.74 m)