Learn More
Electron Microscopy Sciences EMS Style 2A Carbon PVDF Replacement Tips, OAL
SDP
Catalog No. 501930871
Encompass_Preferred
Durable self-aligning replacement PVDF tips and screws for replaceable tip tweezers Style 2A. Flat round tips.
Material: Carbon PVDF
Catalog No. 50-193-0871 Supplier Electron Microscopy Sciences Supplier No. 7832559
Durable self-aligning replacement PVDF tips and screws for replaceable tip tweezers Style 2A. Flat round tips.
Material: Carbon PVDF