Learn More
MSC AbilityOne Carbon-Removing Compound, 55 Gallon Drum, pH 11.5
Catalog No. 502837698
Encompass
This carbon-removing compound is designed for use in soak tanks to effectively soften and remove carbon, gum, and other surface contaminants from various items. It is suitable for cleaning automobile pistons, carburetors, and engine assemblies after they have been pre-cleaned with a degreasing solvent. This compound has a pH of 11.5.
  • Softens and removes carbon, gum, and surface contaminants
  • Suitable for automobile pistons, carburetors, and engine assemblies
  • Designed for soak tank applications
  • Features a pH of 11.5
Catalog No. 50-283-7698 Supplier MSC Supplier No. 80423247
This carbon-removing compound is designed for use in soak tanks to effectively soften and remove carbon, gum, and other surface contaminants from various items. It is suitable for cleaning automobile pistons, carburetors, and engine assemblies after they have been pre-cleaned with a degreasing solvent. This compound has a pH of 11.5.
  • Softens and removes carbon, gum, and surface contaminants
  • Suitable for automobile pistons, carburetors, and engine assemblies
  • Designed for soak tank applications
  • Features a pH of 11.5