Learn More
Electron Microscopy Sciences Carbon Rods, ¼" diameter, 12" OAL
SDP
Catalog No. 5024903
Encompass_Preferred
Technical grade has a typical impurity level of 50ppm or greater and may be used in applications of non critical SEM coating applications.
Diameter: 1/4"
OAL: 12"
Catalog No. 50-249-03 Supplier Electron Microscopy Sciences Supplier No. 70212
Technical grade has a typical impurity level of 50ppm or greater and may be used in applications of non critical SEM coating applications.
Diameter: 1/4"
OAL: 12"