Learn More
Electron Microscopy Sciences Carbon (Graphite) Rods Spectro-Grade, 3/16" diameter, 12" OAL
SDP
Catalog No. 50366164
Encompass_Preferred
Carbon Rods (graphite) with an impurity level of 2ppm or less with a maximum level element of each of 1 ppm. Ideal for all TEM, SEM and other critical applications.
Diameter: 3/16"
OAL: 12"
Catalog No. 50-366-164 Supplier Electron Microscopy Sciences Supplier No. 70231
Carbon Rods (graphite) with an impurity level of 2ppm or less with a maximum level element of each of 1 ppm. Ideal for all TEM, SEM and other critical applications.
Diameter: 3/16"
OAL: 12"