Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)

Catalog No. AA40832KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40832KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40832KS Supplier Thermo Scientific Chemicals Supplier No. 040832KS
Add to Cart

missing translation for 'validMainframeMsg'

 

Used as Magnetron sputtering source. The carbon sputtering target was replaced after the de- positions of 20-30 films with several thousand A in thick- ness in mass-separated negative-ion-beam deposition system. Carbon films can also be deposited by magnetron sputtering in which a magnetically confined plasma torus (race track) acts as a distributed source of ions that impact a carbon sputtering target over a significant part of its area. C -ions were produced from a highly pure carbon-sputtering target by cesium ion sputtering in a neutral and ionized alkaline bombardment-type heavy negative ion source. CN negative ions of 0.88 mA were safely obtained by using a carbon sputtering target and nitrogen gas instead of cyanogen for ionization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Quantity 1 Ea.
Solubility Information Insoluble in water.
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Carbon sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
EINECSNumber 231-955-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.