Learn More
OHAUS Carbon Steel Defender 3000 Series Floor Scale
Catalog No. 01920187 Shop All OHAUS Products
Change view
Click to view available options
Model:
i-DF33P2500B1L
i-DF33P2500B1R
i-DF33XW2500B1R
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-920-187 i-DF33XW2500B1R
01-920-192 i-DF33P2500B1L
01-920-194 i-DF33P2500B1R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-920-187 Supplier OHAUS Supplier No. 30837691
Add to Cart
Add to Cart
Defender 3000 Floor Platforms meet dry weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features a bright display that changes color when in check mode. Scale also includes counting and totalization modes.
  • Painted carbon steel treaded decks with four alloy steel load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
  • Platforms available in sizes of 3 x 3 ft. , 4 x 4 ft. , and 5 x 5 ft. and capacities from 2,500 lb. to 5,000 lb
  • Results are clearly shown on the large, bright backlit LCD display.
  • In checkweighing mode, display changes color (yellow/green/red) to alert users when within the target weight range.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1250 kg
Product Type Scale
Display Type LCD
Readability 0.2 kg
Legal for Trade Class III
Model i-DF33XW2500B1R