Learn More
Ted Pella Inc GEM Carbon Steel Blades, box/200
SDP
Supplier:  Ted Pella Inc 1211
Encompass
Best quality 3 facet blade, single edge, aluminum back. Individually protected. Length: 38.9mm (1.53"). Cutting Edge Length: 38.8mm (1.527"). Width: 19.6mm (0.77"). Thickness: 0.229mm (0.009").
Catalog No. NC9007753