Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades, Sklar 10, Box of 100.
SDP
Catalog No. 501094381 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Carbon Steel Surgical Blades, Sklar, #10. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #10 is most commonly used for large, straight incisions. It has a curved cutting edge. This blade is sterile and available in a box of 100.
Catalog No. 50-109-4381 Supplier Sklar Instruments Supplier No. 063010
Disposable Carbon Steel Surgical Blades, Sklar, #10. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #10 is most commonly used for large, straight incisions. It has a curved cutting edge. This blade is sterile and available in a box of 100.