Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades, Sklar 11. Box of 100.
SDP
Catalog No. 501094382 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Carbon Steel Surgical Blades, Sklar, #11. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #11 is most commonly used for puncture incisions or to open an artery. It has an angled cutting edge that leads to an extremely sharp point. Care should be taken to avoid inadvertently passing the blade too deep into the skin.
Catalog No. 50-109-4382 Supplier Sklar Instruments Supplier No. 063011
Disposable Carbon Steel Surgical Blades, Sklar, #11. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #11 is most commonly used for puncture incisions or to open an artery. It has an angled cutting edge that leads to an extremely sharp point. Care should be taken to avoid inadvertently passing the blade too deep into the skin.