Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades, Sklar 12. Sklar Disposable, Sterile, Box of 100.
SDP
Catalog No. 501094383 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Carbon Steel Surgical Blades, Sklar, #12. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. Disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4383 Supplier Sklar Instruments Supplier No. 063012
Disposable Carbon Steel Surgical Blades, Sklar, #12. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. Disposable blades are available in bulk quantities of 100 per box.