Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades, Sklar 23. Sklar Disposable, Sterile, Carbon Steel, BX100.
SDP
Catalog No. 501094388 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Carbon Steel Surgical Blades, Sklar, #23. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #23 is a leaf-shaped blade sharpened along the leading edge. It is commonly used for making long incisions. Disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-109-4388 Supplier Sklar Instruments Supplier No. 063023
Disposable Carbon Steel Surgical Blades, Sklar, #23. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #23 is a leaf-shaped blade sharpened along the leading edge. It is commonly used for making long incisions. Disposable blades are available in bulk quantities of 100 per box.