Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Tetrabromide 99.0+%, TCI America™
SDP

Catalog No. T0038100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0038100G 100 g
T003825G 25 g
T0038500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. T0038100G Supplier TCI America Supplier No. T0038100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 558-13-4
Molecular Formula CBr4
Molecular Weight (g/mol) 331.627
MDL Number MFCD00000117
InChI Key HJUGFYREWKUQJT-UHFFFAOYSA-N
Synonym carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech
PubChem CID 11205
ChEBI CHEBI:47875
IUPAC Name tetrabromomethane
SMILES C(Br)(Br)(Br)Br

Specifications

Melting Point 94°C
Color White
Boiling Point 190°C
Quantity 100 g
UN Number 2516
Formula Weight 331.63
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Carbon Tetrabromide
EINECSNumber (2)-0041
RTECSNumber FG4725000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.