Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon tetrachloride, 99.8%, for analysis, Thermo Scientific™

Catalog No. AC258530010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC258530010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC258530010 Supplier Thermo Scientific Chemicals Supplier No. 258530010
  • $402.00
    $341.65 / Each of 1

    Save $60.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-23-5
Molecular Formula CCl4
Molecular Weight (g/mol) 153.81
MDL Number MFCD00000785
InChI Key VZGDMQKNWNREIO-UHFFFAOYSA-N
Synonym carbon tetrachloride, perchloromethane, tetrasol, methane, tetrachloro, vermoestricid, benzinoform, necatorine, tetrafinol, carbona, univerm
PubChem CID 5943
ChEBI CHEBI:27385
IUPAC Name tetrachloromethane
SMILES ClC(Cl)(Cl)Cl

Specifications

Decomposition Information Decomposes information: >100°C
Density 1.594g/mL
Assay Percent Range 99.80%
Linear Formula CCl4
Quantity 1 L
Beilstein 01, 64
Fieser 03,43; 04,69; 12,551; 14,72; 16,367
Merck Index 15, 1816
Formula Weight 153.82
Grade Analysis
Chemical Name or Material Carbon tetrachloride
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic if swallowed.
Harms public health and the environment by de
Precautionary Statement Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Toxic: danger of serious damage to health by prolonged exposure through inhalation. Suspected human carcinogen. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Harms public health and the environment by destroying ozone in the upper atmosphere.
NFPA
Health:3 Flammability:0 Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.