Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon tetrachloride, ≥99.95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020485 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0485 1 mL
11-101-9135 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0485 Supplier MilliporeSigma Supelco Supplier No. 026711ML
Add to Cart
Add to Cart
Carbon tetrachloride (CCl4) is a clear, colourless, volatile and very stable chlorinated hydrocarbon. It is widely used as a solvent for fats, varnishes, oils, lacquers, resins, and rubber waxes. It is also used as a dry-cleaning agent, as a refrigerant, and as a starting material in the manufacturing of organic compounds. Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-23-5
Vapor Pressure 143 mmHg (30°C)
Vapor Density 5.32 (vs. air)
Density 1.594 g/mL (at 25°C)
Linear Formula CCl4
MDL Number MFCD00000785
Melting Point -23°C
Boiling Point 76°C to 77°C
Synonym Tetrachloromethane
Type Analytical Standard
Quantity 1 mL
Formula Weight 153.82
Percent Purity ≥99.95% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.