Learn More
Parker Hannifan Parker Hannifin 75344 Activated Carbon Tower, Hydrocarbon Removal, Compatible with N2 Generators, 6 lbs
SDP
Supplier:  Parker Hannifan 75344
Encompass
The Parker Balston Activated Carbon Tower is an essential component for Parker Balston Nitrogen Generators. It effectively removes hydrocarbons from the compressed air supply, ensuring high-purity nitrogen production and protecting membrane integrity. Although it has been marked as discontinued by the manufacturer, its availability remains through various distributors, reinforcing its significance in maintaining generator performance.
Catalog No. NC1852943