Learn More
Ted Pella Inc Carbon Type-B, 200 mesh, Copper, 25/pk
SDP
Catalog No. NC9234417
Encompass
A Formvar film coated with a “heavier” layer of carbon. This is the strongest and most versatile support film we produce. It is stable in the EM under all operating conditions including high magnification with high beam intensity. The films can withstand vigorous specimen preparation techniques. If the carbon surface is hydrophobic, specimen suspensions can be applied to the Formvar surface.
Catalog No. NC9234417 Supplier Ted Pella Inc Supplier No. 01810
A Formvar film coated with a “heavier” layer of carbon. This is the strongest and most versatile support film we produce. It is stable in the EM under all operating conditions including high magnification with high beam intensity. The films can withstand vigorous specimen preparation techniques. If the carbon surface is hydrophobic, specimen suspensions can be applied to the Formvar surface.