Learn More
World Precision Instrument Carbon wire, 49 AWG, 5ft
SDP
Catalog No. 50822140
Encompass
Carbon wire is a single 30-micron fiber of electrochemically activated carbon. This fiber is especially useful in microelectrochemical experiments.
Catalog No. 50-822-140 Supplier World Precision Instrument Supplier No. C3005
Carbon wire is a single 30-micron fiber of electrochemically activated carbon. This fiber is especially useful in microelectrochemical experiments.