Learn More
Abcam 5(6)-Carbo x ynaphthofluorescein, 1MG
Catalog No. 50996234
Encompass_Preferred
MW 952.9 Da, Purity >90%. pH-dependent fluorophore (pKa=7.6) with red fluorescence at pH 9 (Ex/Em: 598/668 nm), fluorescence shifs to shorter wavelengths (Ex/Em: 512/567nm) at neutral or acidic pH environments.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-234 Supplier Abcam Supplier No. AB2755431MG
MW 952.9 Da, Purity >90%. pH-dependent fluorophore (pKa=7.6) with red fluorescence at pH 9 (Ex/Em: 598/668 nm), fluorescence shifs to shorter wavelengths (Ex/Em: 512/567nm) at neutral or acidic pH environments.
The product is subject to the following: Abcam Restricted Use Statement