Learn More
Bioworld Carbonate Buffer, 0.5M, pH 9.6, 500 mL
Catalog No. 50255745
Encompass_Preferred
Product DescriptionCarbonate buffer has a concentration of 0.5M and a pH at 9.6. The pH is determined by the ratio of acid to salt. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-745 Supplier Bioworld Supplier No. 403200232
Product DescriptionCarbonate buffer has a concentration of 0.5M and a pH at 9.6. The pH is determined by the ratio of acid to salt. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.