Learn More
Biotang Inc Carbonate Buffered Saline 5X, pH 9.0. 125 mM Sodium Carbonate, 750 mM Sodium Chloride, pH 9.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-530.
Shop All BIOTANG Inc Products