Learn More
Biotang Inc Carbonate Buffered Saline 5X, pH 9.5. 15 mM Sodium Carbonate, 750 mM Sodium Chloride, pH 9.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-535
Catalog No. 50155802 Shop All BIOTANG Inc Products
Encompass_Preferred
Carbonate Buffered Saline 5X, pH 9.5. 15 mM Sodium Carbonate, 750 mM Sodium Chloride, pH 9.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-535.
Catalog No. 50-155-802 Supplier Biotang Inc Supplier No. BTBB453
Carbonate Buffered Saline 5X, pH 9.5. 15 mM Sodium Carbonate, 750 mM Sodium Chloride, pH 9.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-535.