Learn More
Bioworld Carbonate Buffer, 0.05M, pH 9.4, 1 L
Catalog No. 50255735
Encompass_Preferred
Product DescriptionCarbonate buffer has a concentration of 0.05M and a pH at 9.4.The pH is determined by the ratio of acid to salt. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-735 Supplier Bioworld Supplier No. 403200162
Product DescriptionCarbonate buffer has a concentration of 0.05M and a pH at 9.4.The pH is determined by the ratio of acid to salt. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.