Learn More
Bioworld Carbonate-Buffered Saline, pH 9.0 (5X), 250 mL
Catalog No. 50255750
Encompass_Preferred
Product DescriptionCarbonate-buffered saline contains 125mM sodium carbonate and 750mM sodium chloride, at a pH 9.0. This buffer has a concentration of 5X. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-750 Supplier Bioworld Supplier No. 403200261
Add to Cart
Add to Cart
Product DescriptionCarbonate-buffered saline contains 125mM sodium carbonate and 750mM sodium chloride, at a pH 9.0. This buffer has a concentration of 5X. Carbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.