Learn More
AdipoGen Carbonate ionophore VII
SDP
Catalog No. 502561466
Encompass_Preferred
Chemical. CAS 222310-82-9. Formula C58H79F6NO7. MW 1016.24. Synthetic. Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.
Catalog No. 50-256-1466 Supplier AdipoGen Supplier No. CDXC0131M050
Chemical. CAS 222310-82-9. Formula C58H79F6NO7. MW 1016.24. Synthetic. Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.