Learn More
Medchemexpress LLC Carbonic acid, 2,5-dioxo-1-pyrrolidinyl 1-[5-methoxy-2-nitro-4-[(4-oxo-8,11,14,17-tetraoxa-5-aza... | 1802907-98-7 | 95.0% | 653.63 g/mol | C29H39N3O14 | 50 MG
SDP
Catalog No. 5000203777
Encompass_Preferred
PC Alkyne-PEG4-NHS ester is a cleavable 4-unit PEG linker containing an alkyne group and an N-hydroxysuccinimide (NHS) ester, used for bioconjugation and click-chemistry applications such as antibody-drug conjugate synthesis.
  • Cleavable 4-unit polyethylene glycol spacer for improved solubility.
  • Alkyne functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • NHS ester reactive toward primary amines for efficient conjugation.
  • Typical purity 95.0%.
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-03777 Supplier Medchemexpress LLC Supplier No. HY14013950MG
PC Alkyne-PEG4-NHS ester is a cleavable 4-unit PEG linker containing an alkyne group and an N-hydroxysuccinimide (NHS) ester, used for bioconjugation and click-chemistry applications such as antibody-drug conjugate synthesis.
  • Cleavable 4-unit polyethylene glycol spacer for improved solubility.
  • Alkyne functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • NHS ester reactive toward primary amines for efficient conjugation.
  • Typical purity 95.0%.
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).