Learn More
Medchemexpress LLC Carbonic acid, bicyclo[2.2.1]hept-5-en-2-yl (2,5-dioxo-1-pyrrolidinyl) ester | 1888335-48-5 | 92.1% | 251.24 g/mol | C12H13NO5 | 10 MG
SDP
Catalog No. 5000286309
Encompass_Preferred
Norbornene-NHS is a click-chemistry reagent that functions as a norbornene-derived NHS carbonate dienophile for inverse electron-demand Diels-Alder (IEDDA) conjugation with tetrazines. It is provided for research use to enable efficient amine bioconjugation and labeling applications.
  • Dienophile for inverse electron-demand Diels-Alder (IEDDA) conjugation with tetrazines.
  • NHS carbonate functional group enables efficient coupling to primary amines.
  • Available in small quantities suitable for labeling and bioconjugation experiments.
  • Typical purity about 92.1% and molecular weight 251.24 g/mol.
  • Store sealed and away from moisture; solid at -20°C; in solvent: -80°C for up to 6 months, -20°C for 1 month.
  • Supplied for research use only.
Catalog No. 50-002-86309 Supplier Medchemexpress LLC Supplier No. HY15172710MG
Norbornene-NHS is a click-chemistry reagent that functions as a norbornene-derived NHS carbonate dienophile for inverse electron-demand Diels-Alder (IEDDA) conjugation with tetrazines. It is provided for research use to enable efficient amine bioconjugation and labeling applications.
  • Dienophile for inverse electron-demand Diels-Alder (IEDDA) conjugation with tetrazines.
  • NHS carbonate functional group enables efficient coupling to primary amines.
  • Available in small quantities suitable for labeling and bioconjugation experiments.
  • Typical purity about 92.1% and molecular weight 251.24 g/mol.
  • Store sealed and away from moisture; solid at -20°C; in solvent: -80°C for up to 6 months, -20°C for 1 month.
  • Supplied for research use only.