Learn More
Medchemexpress LLC Carbonic acid, sodium salt (2:3), dihydrate | 6106-20-3 | MFCD00036143 | 226.05 g/mol | Na3H(CO3)2·2H2O | 10g
SDP
Catalog No. 5000268398
Encompass_Preferred
Sodium sesquicarbonate hydrate is a kind of buffering agent that can stabilize pH value by neutralizing acidic substances without causing a sharp change in pH value This property makes it widely used in many fields especially in maintaining acid-base balance in organisms
Catalog No. 50-002-68398 Supplier Medchemexpress LLC Supplier No. HYW92327710G
Sodium sesquicarbonate hydrate is a kind of buffering agent that can stabilize pH value by neutralizing acidic substances without causing a sharp change in pH value This property makes it widely used in many fields especially in maintaining acid-base balance in organisms