Learn More
Medchemexpress LLC Carbonic Anhydrase 1 100ug
SDP
Catalog No. 5000182926
Encompass_Preferred
Carbonic Anhydrase 14 Protein catalyzes the reversible hydration of carbon dioxide converting it to bicarbonate ions and protons and vice versa Its vital enzymatic activity contributes to physiological processes regulating acid-base balance respiration and cellular pH homeostasis The protein plays a pivotal role in carbon dioxide transport and metabolism in the body Carbonic Anhydrase 14 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 14 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-82926 Supplier Medchemexpress LLC Supplier No. HYP72867100UG
Carbonic Anhydrase 14 Protein catalyzes the reversible hydration of carbon dioxide converting it to bicarbonate ions and protons and vice versa Its vital enzymatic activity contributes to physiological processes regulating acid-base balance respiration and cellular pH homeostasis The protein plays a pivotal role in carbon dioxide transport and metabolism in the body Carbonic Anhydrase 14 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 14 protein expressed by HEK293 with C-His labeled tag