Learn More
Medchemexpress LLC Carbonic Anhydrase 1 5ug
SDP
Catalog No. 5000181592
Encompass_Preferred
Carbonic Anhydrase 10 Protein a member of the carbonic anhydrase family plays a role in the regulation of pH and ion transport It is expressed in various tissues including the brain liver and kidney The potential involvement of Carbonic Anhydrase 10 Protein in diseases and its physiological functions make it a subject of interest in biomedical research Carbonic Anhydrase 10 Protein Mouse (HEK293 His) is the recombinant mouse-derived Carbonic Anhydrase 10 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81592 Supplier Medchemexpress LLC Supplier No. HYP728655UG
Carbonic Anhydrase 10 Protein a member of the carbonic anhydrase family plays a role in the regulation of pH and ion transport It is expressed in various tissues including the brain liver and kidney The potential involvement of Carbonic Anhydrase 10 Protein in diseases and its physiological functions make it a subject of interest in biomedical research Carbonic Anhydrase 10 Protein Mouse (HEK293 His) is the recombinant mouse-derived Carbonic Anhydrase 10 protein expressed by HEK293 with C-His labeled tag