Learn More
Medchemexpress LLC Carbonic Anhydrase 2 100ug | 100ug
SDP
Catalog No. 5000194993
Encompass_Preferred
Carbonic Anhydrase 2 (CA2) catalyzes the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon Two transcript variants encoding different isoforms have been identified Carbonic Anhydrase 2 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94993 Supplier Medchemexpress LLC Supplier No. HYP78746100UG
Carbonic Anhydrase 2 (CA2) catalyzes the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon Two transcript variants encoding different isoforms have been identified Carbonic Anhydrase 2 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 2 protein expressed by HEK293 with C-His labeled tag