Learn More
Medchemexpress LLC Carbonic Anhydrase 2 500ug
SDP
Catalog No. 5000181647
Encompass_Preferred
Carbonic Anhydrase 2 (CA2) catalyzes the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon Two transcript variants encoding different isoforms have been identified Carbonic Anhydrase 2 Protein Human (C-His) is the recombinant human-derived Carbonic Anhydrase 2 protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-81647 Supplier Medchemexpress LLC Supplier No. HYP72860A500UG
Carbonic Anhydrase 2 (CA2) catalyzes the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon Two transcript variants encoding different isoforms have been identified Carbonic Anhydrase 2 Protein Human (C-His) is the recombinant human-derived Carbonic Anhydrase 2 protein expressed by E coli with C-6 His labeled tag