Learn More
Crescent Chemical Co Inc CARBONIC ANHYDRASE
SDP
Catalog No. 50247397
Encompass_Preferred
Carbonic anhydrase from bovine erythrocytes ca. 1.5 U/mg lyophil. Homogeneous in SDS-PAGE. Contains carbonic anhydrases A and B;both forms have similar high specific activities and therefore belong to the C-group of mammalian carbonic anhydrases (1). Size - 100MG Storage Conditions - -15 °C TO
Catalog No. 50-247-397 Supplier Crescent Chemical Co Inc Supplier No. SE15882.02
Carbonic anhydrase from bovine erythrocytes ca. 1.5 U/mg lyophil. Homogeneous in SDS-PAGE. Contains carbonic anhydrases A and B;both forms have similar high specific activities and therefore belong to the C-group of mammalian carbonic anhydrases (1). Size - 100MG Storage Conditions - -15 °C TO