Learn More
Medchemexpress LLC Carbonic Anhydrase 4 5ug
SDP
Catalog No. 5000181871
Encompass_Preferred
Carbonic anhydrase 4 catalyzes the reversible hydration of carbon dioxide and plays a crucial role in maintaining intracellular and extracellular pH Carbonic Anhydrase 4 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81871 Supplier Medchemexpress LLC Supplier No. HYP728625UG
Carbonic anhydrase 4 catalyzes the reversible hydration of carbon dioxide and plays a crucial role in maintaining intracellular and extracellular pH Carbonic Anhydrase 4 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 4 protein expressed by HEK293 with C-His labeled tag