Learn More
Medchemexpress LLC Carbonic Anhydrase 9 100ug
SDP
Catalog No. 5000182683
Encompass_Preferred
The carbonic anhydrase 9 (CA9) protein plays a crucial role as a catalyst in the conversion of carbon dioxide and water It promotes the formation of bicarbonate and hydrogen ions (dissociated ions of carbonic acid) Carbonic Anhydrase 9 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived Carbonic Anhydrase 9 protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-82683 Supplier Medchemexpress LLC Supplier No. HYP72348100UG
The carbonic anhydrase 9 (CA9) protein plays a crucial role as a catalyst in the conversion of carbon dioxide and water It promotes the formation of bicarbonate and hydrogen ions (dissociated ions of carbonic acid) Carbonic Anhydrase 9 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived Carbonic Anhydrase 9 protein expressed by HEK293 with C-Avi C-6 His labeled tag