Learn More
Medchemexpress LLC Recombinant human carbonic anhydrase 9 (His-tag) | >95.0% | 100UG
SDP
Catalog No. 5000177971
Encompass_Preferred
Recombinant human carbonic anhydrase 9 (CA9) produced in HEK293 cells and supplied as a lyophilized preparation with a C-terminal 6xHis tag. The protein corresponds to amino acids Q38-D414 of human CA9, is glycosylated (apparent molecular weight 45-60 kDa), and is purified to >95% by reducing SDS-PAGE. It is formulated in PBS (pH 7.4) or PBS with 8% trehalose and intended for research use in biochemical and cell biology studies involving CA9.
Catalog No. 50-001-77971 Supplier Medchemexpress LLC Supplier No. HYP70064100UG
Recombinant human carbonic anhydrase 9 (CA9) produced in HEK293 cells and supplied as a lyophilized preparation with a C-terminal 6xHis tag. The protein corresponds to amino acids Q38-D414 of human CA9, is glycosylated (apparent molecular weight 45-60 kDa), and is purified to >95% by reducing SDS-PAGE. It is formulated in PBS (pH 7.4) or PBS with 8% trehalose and intended for research use in biochemical and cell biology studies involving CA9.