Learn More
Medchemexpress LLC Carbonic anhydrase 9 protein, cynomolgus (HEK293, His) | 10UG
SDP
Catalog No. 5000176164
Encompass_Preferred
Recombinant cynomolgus carbonic anhydrase IX (CA9) expressed in HEK293 cells with a C-terminal His tag. The enzyme catalyzes reversible hydration of CO2 and is supplied as a lyophilized powder from PBS (pH 7.4) with 8% trehalose; reconstitute to ≥100 μg/mL in ddH2O for recommended use. Suitable for biochemical and cell biology studies of pH regulation and CA9-related pathways.
  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose for stability.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Useful for studies of carbon dioxide transport and pH regulation.
Catalog No. 50-001-76164 Supplier Medchemexpress LLC Supplier No. HYP70088210UG
Recombinant cynomolgus carbonic anhydrase IX (CA9) expressed in HEK293 cells with a C-terminal His tag. The enzyme catalyzes reversible hydration of CO2 and is supplied as a lyophilized powder from PBS (pH 7.4) with 8% trehalose; reconstitute to ≥100 μg/mL in ddH2O for recommended use. Suitable for biochemical and cell biology studies of pH regulation and CA9-related pathways.
  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose for stability.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Useful for studies of carbon dioxide transport and pH regulation.