Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbonic anhydrase 9 protein (cynomolgus) | >95.0% | 50UG
SDP

Catalog No. 5000177266
Encompass_Preferred

Recombinant cynomolgus carbonic anhydrase 9 (CA9) expressed in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein. The product is provided as a sterile powder formulated in PBS with 8% trehalose, characterized by greater than 95% purity and a molecular weight of about 50-60 kDa. It is intended for research on carbonic anhydrase activity, pH regulation, and tumor biology.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag enables detection and purification.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Sequence range Q38-D402 corresponding to the mature protein.
  • Lyophilized sterile powder formulated in PBS with 8% trehalose.
  • Suggested reconstitution at ≥100 μg/mL in ddH2O.
  • Store at -20°C for up to 2 years; aliquot and freeze at -80°C for extended storage.

Catalog No. 50-001-77266 Supplier Medchemexpress LLC Supplier No. HYP70088250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant cynomolgus carbonic anhydrase 9 (CA9) expressed in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein. The product is provided as a sterile powder formulated in PBS with 8% trehalose, characterized by greater than 95% purity and a molecular weight of about 50-60 kDa. It is intended for research on carbonic anhydrase activity, pH regulation, and tumor biology.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag enables detection and purification.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Sequence range Q38-D402 corresponding to the mature protein.
  • Lyophilized sterile powder formulated in PBS with 8% trehalose.
  • Suggested reconstitution at ≥100 μg/mL in ddH2O.
  • Store at -20°C for up to 2 years; aliquot and freeze at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.