Learn More
Medchemexpress LLC Carbonic anhydrase 9 protein, canine (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000291368
Encompass_Preferred
Recombinant canine Carbonic Anhydrase 9 (CA IX) expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use, this reagent is intended for antibody binding assays and biochemical characterization. Formulated in PBS (pH 7.4) and provided in microgram-scale quantities.
Catalog No. 50-002-91368 Supplier Medchemexpress LLC Supplier No. HYP7545410UG
Recombinant canine Carbonic Anhydrase 9 (CA IX) expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use, this reagent is intended for antibody binding assays and biochemical characterization. Formulated in PBS (pH 7.4) and provided in microgram-scale quantities.