Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbonic anhydrase, bovine erythrocytes | 9001-03-0 | 29.0 kDa | 10 MG
SDP

Catalog No. 5000424159
Encompass_Preferred

Carbonic anhydrase, bovine erythrocytes (EC 4.2.1.1) is a ubiquitous zinc-containing metalloenzyme found in prokaryotes and eukaryotes. It catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This enzyme can be utilized for research related to cancer, glaucoma, obesity, and epilepsy. For in vitro use, the lyophilized powder should be dissolved in ice-cold water at a concentration of 0.1 mg/mL and stored in an ice bath until use.

  • Molecular weight: 29.0 kDa
  • Optimal pH: 7.0-7.5
  • Isoelectric point: 6.40
  • Inhibitors include monovalent anions, sulfonates, sulfonamides, and imidazole
  • Enzyme activity: ≥3000 U/mg solid
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: Soluble in water at ≥ 50 mg/mL

Catalog No. 50-004-24159 Supplier Medchemexpress LLC Supplier No. HYP177510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbonic anhydrase, bovine erythrocytes (EC 4.2.1.1) is a ubiquitous zinc-containing metalloenzyme found in prokaryotes and eukaryotes. It catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This enzyme can be utilized for research related to cancer, glaucoma, obesity, and epilepsy. For in vitro use, the lyophilized powder should be dissolved in ice-cold water at a concentration of 0.1 mg/mL and stored in an ice bath until use.

  • Molecular weight: 29.0 kDa
  • Optimal pH: 7.0-7.5
  • Isoelectric point: 6.40
  • Inhibitors include monovalent anions, sulfonates, sulfonamides, and imidazole
  • Enzyme activity: ≥3000 U/mg solid
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: Soluble in water at ≥ 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.