Learn More
Sino Biological Carbonic anhydrase II Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 201408T40100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Carbonic anhydrase II fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-8193