Learn More
Medchemexpress LLC Carbonic anhydrase inhibitor 2 | 2758231-43-3 | 98.4% | C12H16N4O6S | 10MG
SDP
Catalog No. 5000261018
Encompass_Preferred
Carbonic anhydrase inhibitor 2 is a research-stage small molecule characterized as a carbonic anhydrase II inhibitor (referred to as compound 7c in the originating study). It is supplied as a white to off-white solid for laboratory research use and has reported high purity and defined storage recommendations.
  • Inhibits carbonic anhydrase II and reduced intraocular pressure in glaucomatous rabbits.
  • White to off-white solid suitable for research applications.
  • High purity, 98.4% by supplier specification.
  • Molecular formula C12H16N4O6S and molecular weight 344.34.
  • Stable under recommended storage: powder stored at -20°C for long-term.
Catalog No. 50-002-61018 Supplier Medchemexpress LLC Supplier No. HY14284910MG
Carbonic anhydrase inhibitor 2 is a research-stage small molecule characterized as a carbonic anhydrase II inhibitor (referred to as compound 7c in the originating study). It is supplied as a white to off-white solid for laboratory research use and has reported high purity and defined storage recommendations.
  • Inhibits carbonic anhydrase II and reduced intraocular pressure in glaucomatous rabbits.
  • White to off-white solid suitable for research applications.
  • High purity, 98.4% by supplier specification.
  • Molecular formula C12H16N4O6S and molecular weight 344.34.
  • Stable under recommended storage: powder stored at -20°C for long-term.