Learn More
Medchemexpress LLC Carbonic anhydrase (isoenzyme) | 9001-03-0 | 110000.00 | 25 MG
SDP
Catalog No. 5000426691
Encompass_Preferred
Carbonic anhydrase isoenzyme is a ubiquitous zinc-containing metalloenzyme found in prokaryotes and eukaryotes. It is an isoenzyme of Carbonic anhydrase (HY-P1775) and catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This enzyme is utilized in research related to cancer, glaucoma, obesity, and epilepsy.
  • Ubiquitous zinc-containing metalloenzyme present in prokaryotes and eukaryotes.
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons.
  • Useful for research in areas such as cancer, glaucoma, obesity, and epilepsy.
  • Available in various quantities: 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
Catalog No. 50-004-26691 Supplier Medchemexpress LLC Supplier No. 25MGHYP1775A
Carbonic anhydrase isoenzyme is a ubiquitous zinc-containing metalloenzyme found in prokaryotes and eukaryotes. It is an isoenzyme of Carbonic anhydrase (HY-P1775) and catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This enzyme is utilized in research related to cancer, glaucoma, obesity, and epilepsy.
  • Ubiquitous zinc-containing metalloenzyme present in prokaryotes and eukaryotes.
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons.
  • Useful for research in areas such as cancer, glaucoma, obesity, and epilepsy.
  • Available in various quantities: 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.