Learn More
Medchemexpress LLC Carbonic anhydrase (isoenzyme) | 9001-03-0 | ≥3000 U/mg solid | 110000.00 | 5 MG
SDP
Catalog No. 5000422731
Encompass_Preferred
Carbonic anhydrase isoenzyme is a ubiquitous zinc-containing metalloenzyme that catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This product can be used for research in various fields including cancer, glaucoma, obesity, and epilepsy.
  • Ubiquitous zinc-containing metalloenzyme
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons
  • Suitable for research in cancer, glaucoma, obesity, and epilepsy
Catalog No. 50-004-22731 Supplier Medchemexpress LLC Supplier No. HYP1775A5MG
Carbonic anhydrase isoenzyme is a ubiquitous zinc-containing metalloenzyme that catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This product can be used for research in various fields including cancer, glaucoma, obesity, and epilepsy.
  • Ubiquitous zinc-containing metalloenzyme
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons
  • Suitable for research in cancer, glaucoma, obesity, and epilepsy