Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbonic Anhydrase Isozyme II from bovine erythrocytes lyophilized powder, ≥3,000 W-A units/mg protein | 9001-03-0 | MFCD00081467 | 5mg
Catalog No. 501785459
Encompass
Carbonic Anhydrase Isozyme II from bovine erythrocytes lyophilized powder, ≥3,000 W-A units/mg protein | 9001-03-0 | MFCD00081467 | 5mg
Catalog No. 501785459 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C25225MG
Carbonic Anhydrase Isozyme II from bovine erythrocytes lyophilized powder, ≥3,000 W-A units/mg protein | 9001-03-0 | MFCD00081467 | 5mg