Learn More
Biomatik Corporation Carbonic Anhydrase Iv (Ca4) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29559
Encompass_Preferred
Carbonic Anhydrase Iv (Ca4) Monoclonal Antibody is a Rat (Rattus norvegicus) Monoclonal Antibody to Carbonic Anhydrase IV (CA4). The Carbonic Anhydrase Iv (Ca4) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Rat (Rattus norvegicus). Immunogen: Ser20~Ser220. Applications: WB. Target Synonyms: Carbonic Anhydrase IV (CA4). Accession Number: P48284. Gene ID: 29242. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5331