Learn More
ACROBiosystems Carbonic Anhydrase IX 25tests
Catalog No. 502535433
Encompass_Preferred
APC-Labeled Human Carbonic Anhydrase IX (38-414) Protein His Tag (CA9-HA2H7) is produced via conjugation of APC to Human Carbonic Anhydrase IX (38-414) Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human Carbonic Anhydrase IX (38-414) Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).
Catalog No. 50-253-5433 Supplier ACROBiosystems Supplier No. CA9HA2H725TESTS
APC-Labeled Human Carbonic Anhydrase IX (38-414) Protein His Tag (CA9-HA2H7) is produced via conjugation of APC to Human Carbonic Anhydrase IX (38-414) Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human Carbonic Anhydrase IX (38-414) Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).