Learn More
ACROBiosystems FITC-Labeled Human Carbonic Anhydrase IX (38-414), His Tag (CA9-HF228) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).
Catalog No. 502231473
Encompass_Preferred
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.7 kDa.
This item is not returnable. View return policy
Catalog No. 50-223-1473 Supplier ACROBiosystems Supplier No. CA9HF22825UG
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.7 kDa.