Learn More
ACROBiosystems Recombinant Protein;Human Carbonic Anhydrase II / CA2 Protein;HEK293;1MG;CA2,CA2,CA-II,CAII,Car2,CA-2,Carbonic Anhydrase II
Catalog No. 501658991
Encompass_Preferred
Recombinant Protein;Human Carbonic Anhydrase II / CA2 Protein;1MG;Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Lys 260 (Accession # NP_000058.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8991 Supplier ACROBiosystems Supplier No. CA2H52281MG
Recombinant Protein;Human Carbonic Anhydrase II / CA2 Protein;1MG;Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Lys 260 (Accession # NP_000058.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE).